Would you like to log in again?
Innovating Science® Acids and Bases Lab Activity
Supplier: ALDON CORP SE
Innovating Science®
IS1500 IS1001
470328-340KT 19.8 USD InStock
470328-340 470163-306
Innovating Science® Acids and Bases Lab Activity
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
In this activity, students make hypotheses regarding household items and then test them.
  • Perfect Introduction to Acids and Bases
  • Students Test Household Items
  • Kit Features Extra Litmus Paper for More Experiments
  • All Required Chemicals are Included
  • Acid and Base Chemistry in Microscale
Students learn about acids and bases through a class discussion and then test a dilute acid and dilute base with neutral litmus paper to learn how the paper indicates something is acidic or basic.
Once students master that skill, they can move on to test some common household items to determine if they are acids or bases. Extra litmus paper is included to test more items either from home or around the school building for a greater understanding of acids and bases. 
Ordering information: This kit includes hydrochloric acid solution, sodium hydroxide solution, distilled water, vinegar, dilute ammonia, liquid soap, lemon juice, neutral litmus paper, spot plates, a teacher's guide, and a student copy master.
Delivery information: Instructor's manual and SDS included.
Caution: Caution:CHOKING HAZARD This item has small parts and is not for children under three years.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty