Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Forensics of Soil and Mineral Analysis
Supplier: ALDON CORP SE
IS9015 IS9015-REFILL
470163-320KT 224 USD InStock
470163-320 470163-318
Forensics of Soil and Mineral Analysis
Educational Materials Earth Science Educational Materials Minerals and Rocks Learning Activities
A Crime Has Been Reported and It's Up to You to Help Prove Who Did It!
  • Forensic physical chemistry experiment
  • All chemicals required are included
This kit has been designed to introduce students to the field of forensic soil analysis by providing them with a set of soil and glass samples that have been recovered from a crime scene to compare with a set of samples taken from various suspects. Students will compare samples based on physical observations using polarized, UV and standard light both under ambient conditions and after heating the samples. The properties of the samples will be tested by measuring particle size distribution, material acidity, particle density gradient and mineral composition. This kit contains 10 unique sets of materials that will accommodate up to 30 students when working in groups of 3.
Ordering information: Includes Universal Indicator, Bromoform, Bromobenzene, Potassium Chromate Solution, Hydrochloric Acid Solution, Soil Sample A, Soil Sample B, Soil Sample C, Soil Sample D, Crime Scene Soil Sample, Glass Sample A, Glass Sample B, Crime Scene Glass Sample, Magnifying Glass, Forceps, Capped Test Tubes, Polarizing Filters, Drying Dishes, Sieve Screen; Small, Sieve Screen; Large, Glass Slides, Plastic Scoops, and Teacher's Guide and Student Copymaster.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty