Would you like to log in again?
Diaphragm with 3 Double Slits of Different Widths
Supplier: AMERICAN 3B SCIENTIFIC SE
1000596
470163-768EA 28.35 USD InStock
470163-768
Diaphragm with 3 Double Slits of Different Widths
Educational Materials Physics Educational Materials Optics Learning Activities
Photographic diffraction objects in a slide frame.
  • Dimensions: 50x50 mm²
  • Number of slits: 3 sets of 2
  • Slit width [mm]: 0.10; 0.15; 0.20
  • Slit spacing [mm]: 0.30
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty