Would you like to log in again?
Singing Rods
Supplier: AMERICAN SCIENTIFIC LLC SE
3637-00
470163-896EA 24.2 USD InStock
470163-896
Singing Rods
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
A must for explorations of sound and waves.
  • Sold as a set of two
  • Approx length: 20" (500mm) and 30" (750mm)
  • Comes with a Teacher's Guide with detailed instructions and a bag of crushed rosin to lightly coat your fingertips
These rods are an easy way to demonstrate longitudinal waves as opposed to transverse waves. They're ideal for teaching about nodes and anti-nodes in standing waves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty