Would you like to log in again?
Revolution
Supplier: AMERICAN SCIENTIFIC LLC SE
3164-00
470163-900EA 13.85 USD InStock
470163-900
Revolution
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A Fascinating Demonstration!
  • Approximate product size: 2-1/2" x 5" x 3" deep
  • Batteries are not required
  • Teacher's Guide included
This fascinating anti-gravity demonstration works on the principle of repelling magnets and is ideal for discussing magnet levitation and frictional force experiments. With a simple twist of the fingers the axle and spins horizontally in a state of near perpetual motion for up to 20 minutes. Black and white reflecting strips on axle allow students to observe the patterns created by the alternating colors and to measure rotation speed using a stroboscope (not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty