Would you like to log in again?
Harbottle
Supplier: AMERICAN SCIENTIFIC LLC SE
6080-40
470163-910EA 47.5 USD InStock
470163-910
Harbottle
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Excellent teaching tool for introducing pressure and discussing its applications such as vacuum cleaners, flight, pumps, weather, and even how we breathe.
  • Teacher's Guide Included
  • Glass Bottle Included
  • Two Balloons Included
  • For Grades Five and Above
  • No Twisting or Tying Required
Place the balloon into the opening of the bottle and stretch the mouth of the balloon over the mouth of the bottle. Blow into the balloon, and when it is fully inflated, close the bottom hole of the bottle with a stopper. The normal air pressure from outside allows the balloon to stay inflated. No twisting or tying required.
Demonstrate the strength of air pressure by filling the balloon with water before releasing the stopper. The Harbottle is designed for grades five and above.
Ordering information: This kit includes a glass bottle, two balloons, rubber stopper, and teacher's guide with background information and activity suggestions for students.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty