Would you like to log in again?
Vacuum Lifter
Supplier: AMERICAN SCIENTIFIC LLC SE
AR-510
470163-920EA 31.25 USD InStock
470163-920
Vacuum Lifter
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Demonstrates Atmospheric Pressure.
  • Explore additional concepts such as surface area, force, vacuums, and pressure
This durable, thick rubber mat has a simple design. Pick up an ordinary flat lab stool or make a challenge to lift off a table, which is nearly impossible! Show how suction cups stay stuck because of the pushing force of air, not from a pulling "suction" force. Comes with instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty