Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Solar Cell with Wires

Supplier: SUNDANCE SOLAR MO
700-10850-14W
470163-932EA 11.6 USD InStock
470163-932
Solar Cell with Wires
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Photocells with included connecting wires

  • High Efficiency Miniature Solar Cell
  • Can easily be arranged for larger load applications including higher voltage and current loads.
  • Miniature compact size with solid attractive casing.

Monocrystalline Solar Cell
Efficiency: 16-17%
Color: Blue
Peak Voltage (Vmp): 6V
Open Circuit Voltage (Voc): 7.2V
Peak Current (Imp): 70mA
Short Circuit Current (Isc): 75mA
Dimensions: 95mm x 51mm x 3mm (LxWxD)

Includes connecting wires

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty