Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Photoconductivity

Supplier: OSAW INDUSTRIAL PRODUCTS SE
SK007
470163-952EA 445.4 USD InStock
470163-952
Photoconductivity
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This apparatus is used to study the current Vs voltage characteristics of CdS photo-resistor at constant irradiance and to measure the photo-current.

Salient features:
Adjustable slit self centering.
Marked analyzer and polarizer for angle measurement.

Principle and Working:
Absorption of light increases electrical conductivity in a solid. In case of cadmium sulphide (CdS) photoresistor, when it is exposed to light the transition of activator electrons into the conduction band and charge exchange of traps with holes being created in the valence band causing enhancement of conductivity. In the present setup, photo current is studied as a function of the voltage applied to the photo resistor at a constant irradiance. The angle of polarization is changed by polarizer filter.

Included:
Photoresistor in holder (CdS)
Lamp housing
Convex lens in holder
Polarizer/Analyzer (pair)
Optical Bench
Adjustable slit self centering
Digital multimeter
Flexible plug lead, 50 cm (Red) 2
Flexible plug lead, 1 m (Black)

Required but not included: Power supply 12V AC/DC 5 Amp. 1 and Regulated Power Supply, 0-16V DC, 5A.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty