Would you like to log in again?
Boyle's Law
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SH302
470163-990CS 130.55 USD InStock
470163-990
Boyle's Law
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This apparatus is used to Verifying the Boyle’s law when air is decompressed and compressed. A horizontally mounted acrylic tube having a graduated scale (0- 300cm3) along one side. A gas tap closes the tube. Pressure gauge is present next to gas tap. By changing the position of rubber stopper, the effective length of the tube can be selected. The scale measures volume and pressure by means of pressure gauze. Boyle’s law is investigated by changing position of the stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty