Would you like to log in again?
Speed of Light Apparatus
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SK077
470163-998EA 518.15 USD InStock
470163-998
Speed of Light Apparatus
Educational Materials Physics Educational Materials Optics Learning Activities
A classical experiment to determine the speed of light in air.
  • 1MHz internal modulation frequency
  • Dual photo detector
Principle and Working:
The LASER beam from the Transmitter strikes the Beam Splitter to split the beam in two perpendicular directions. The first beam is transmitted through Transmitter directly and is received in first channel of the receiver. The second perpendicular beam from Beam Splitter travels a larger distance by being reflected back from a mirror placed about 5 meter or more and is received in second channel of the receiver. In the present setup, the time difference between the two peaks of received LASER signals can be observed on a oscilloscope (40MHz) and can be used to calculate the Speed of Light.
Ordering information: Includes Laser Transmitter Module, Laser Receiver Module, Power Adaptor 5V, Beam Splitter, Convex Lens f=50mm, Cylindrical Base, Reflecting Mirror in Mount, 'A' Base, Support Rod (1m), BNC to BNC Cable. Required but not included: Oscilloscope 40 MHz.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty