Would you like to log in again?
CENCO® Assorted Ball Set
Supplier: VWR International
DBLST12-A
470164-716EA 158.05 USD InStock
470164-716
CENCO® Assorted Ball Set
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Comprehensive and High Quality.
  • Packed in plastic storage box
  • Set of 12
A set with 12 balls in all, includes 25mm diameter drilled balls of brass, aluminum, wood, steel, copper and lead; two 25mm solid steel balls, one plain and one with bifilar suspension lugs; 38 mm balls of brass, steel and lead with hooks; and one 25mm solid cork ball.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty