Would you like to log in again?
Ballistic Car with Remote
Supplier: United Scientific Supplies
BLCAR2
470164-718EA 133.15 USD InStock
470164-718
Ballistic Car with Remote
Educational Materials Physics Educational Materials Mechanics Learning Activities
This Ballistics Cart demonstrates the independence of vertical and horizontal motions.
  • Operating Instructions Included
  • Two Steel Balls Included
  • For Demonstrating the Independence of Vertical and Horizontal Motions
  • Conical Rubber Catcher Included
  • Spring-Loaded Piston
It consists of a four-wheeled cart carrying a vertical tube with a spring-loaded piston inside. The tube is loaded with a ball. When the cart is set in motion, the piston is released remotely by a wireless signal, projecting the ball upwards out of the tube. Both the ball and the cart share the same horizontal velocity, which is unaffected by gravity, so that when the ball falls back down, it lands back in the tube and does not get "left behind" by the cart.
The cart includes a conical rubber catcher to compensate for slight trajectory variations.
Ordering information: This kit includes two 3/4" steel balls and operating instructions. The car requires one 9V battery, not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty