Would you like to log in again?
KidWind Advanced Wind Experiment Kit
Supplier: VERNIER SOFTWARE & TECH SE
KW-AWX
470165-106EA 173.99 USD InStock
470165-106
KidWind Advanced Wind Experiment Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
All you need to add is a wind source, basic tools, and imagination!
  • Create blades for wind turbines
  • Design applications for wind power
  • Grades 7 - 12+
Perform all of the same investigations as the Basic Wind Experiment Kit with the ability to generate more power at higher voltages (0.5 V–20 V). Discover advanced concepts of wind turbine technology, including gearboxes and generator construction (with the GenPack add-on). Students can use the blades they design to generate electricity, lift weights, and pump water. This kit is perfect for grades 7–12 and college.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty