Would you like to log in again?
Chipboard Sheets
Supplier: VERNIER SOFTWARE & TECH SE
KW-CB50
470165-132PK 18.99 USD InStock
470165-132
Chipboard Sheets
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Chipboard is a stiff, easy to cut, lightweight, low cost, and recyclable material used to make simple wind turbine blades.
  • Length: 12"
  • Width: 3"
  • Thickness: 1.5 mm
Chipboard is a great material used to make simple wind turbine blades. It is stiff, easy to cut, lightweight, low cost and recyclable. Each package comes with 50 chipboard sheets that are 3" by 12". Each sheet is approximately 1.5mm thick, so they will stay stiff in a light wind but bend in a strong one.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty