Would you like to log in again?
Eisco® Rectangular Retort Stand, Rod, Clamp & Ring Set
Supplier: EISCO SCIENTIFIC LLC
MTST6
470165-384EA 110.25 USD InStock
470165-384
Eisco® Rectangular Retort Stand, Rod, Clamp & Ring Set
Support Stands
6 piece laboratory set designed for most general science laboratories.
The durable and sturdy construction of this set makes it an excellent choice for science classrooms and laboratories. The (10×9" L×W) base is made of corrosion resistant, powder coated pressed steel and features a tapped hole, which fits the included rod, and can accommodate any rod with a 10×1.5 mm metric thread. The stainless steel rod measures 24" tall and is 12 mm in diameter. The zinc plated support rings measure 4.75" from center to support and include integrated screw clamps. The bosshead features 2 metal thumb screws and is able to stabilize the clamp at a 90 degree angle to the rod. The 3 finger clamp features 2 adjusting thumb screws and is vinyl coated.
Delivery information: Includes:
1× Retort base made of corrosion resistant, powder coated steel
1× Stainless steel rod featuring a 10×1.5 mm metric thread
2× Metal support ring
1× Vinyl coated three-prong adjustable clamp
1× Metal dual adjusting bosshead
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty