Would you like to log in again?
Premium Two-Prong Clamp, Cork Lined
Supplier: EISCO SCIENTIFIC LLC
PRCMP08BH PRCMP08
470165-416EA 20.3 USD InStock
470165-416 470165-414
Premium Two-Prong Clamp, Cork Lined
Clamps General Purpose Clamps
Cork Lined Extension Lab Clamp
  • Maximum clamp opening is 11 cm
  • 14 cm connecting rod.
Clamp constructed of powder coated steel, two cork lined prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty