Would you like to log in again?
Laboratory Retort Stand with Ring Set
Supplier: EISCO SCIENTIFIC LLC
CH0652BR8
470165-432EA 65.45 USD InStock
470165-432
Laboratory Retort Stand with Ring Set
Support Stands
This laboratory set includes a retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10 mm x 1.5 mm metric thread and three cast iron support rings.
  • Stainless steel resists scaling and corrosion
  • Durable and sturdy construction
  • Superb stability for basic experiments and setups
  • Base features a tapped hole
The 8" long by 5" wide base weighs 1.75 lbs, providing superb stability for experiments & setups. The included rod measures 20" tall and is 12mm in diameter. Includes three support rings, with diameters of 3", 4" and 5" to support a variety of apparatus. Each ring features a zinc plated steel shank secured to a screw clamp. Distance from each of the rings centers to the center of the support measures 4.75".
The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratory setups.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty