Would you like to log in again?
Rectangular Retort Stand, Rod, Clamp and Ring Set
Supplier: EISCO SCIENTIFIC LLC
MTST5
470165-450EA 67.9 USD InStock
470165-450
Rectangular Retort Stand, Rod, Clamp and Ring Set
Support Stands
This laboratory set includes a retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10 mm x 1.5 mm metric thread, a support ring and a three-pronged coated clamp.
The 8" long by 5" wide base weighs 1.75 lbs, providing superb stability for experiments & setups. The included rod measures 23.6" tall and is 12 mm in diameter.
Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10 mm x 1.5 mm metric thread.
The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories & basic professional laboratory setups.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty