Would you like to log in again?
Blue Epoxy-Coated Steel Wire Test Tube Racks
Supplier: EISCO SCIENTIFIC LLC
CH182015C CH182015D CH182015B CH182015A CH182015G CH182015E CH182015F CH182015H
470165-448EA 52.75 USD InStock
470165-448 470165-446 470165-444 470165-456 470165-442 470165-454 470165-452 470165-390
Blue Epoxy-Coated Steel Wire Test Tube Racks
Racks Tube Racks
Coated steel wire racks are chemical and corrosion resistant and come in a variety of sizes.
  • Test tube rack featuring a sturdy epoxy-coated steel construction that will stand up to the wear and tear any lab
  • Withstands temperatures up to 121 °C (249.8 °F)
Coating provides a protective and smooth coat that is impervious to weak acids, bases, and salts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty