Would you like to log in again?
Go Direct™ Melt Station
Supplier: VERNIER SOFTWARE & TECH SE
GDX-MLT
470166-514EA 598.99 USD InStock
470166-514
Go Direct™ Melt Station
Data Loggers
The Go Direct Melt Station accurately determines the melting temperature of solid substances.
  • The Go Direct Melt Station can be used in a variety of experiments
  • Determine the melting temperature of an known organic solid such as aspirin, ibuprofen, or caffeine
  • Identify an unknown organic solid by its melting temperature
  • Use melting temperature to analyze reaction products
  • Connects via Bluetooth® wireless technology or via USB to your device
Teach students the visual detection capillary method of melting point determination with the Go Direct Melt Station. It accurately measures melting temperatures of a solid (up to 260°C), and the real-time graphing provides a unique perspective of the melting process.
The wide-angle observation and magnification window, LED-lit heater block, and adjustable tilt base give students a clear view of the substance as they witness the state change. Internal cooling fans reduce the waiting time between sample testing. Also included is an important safety feature that automatically powers down the heating block after 60 minutes with no change to the control knob.
Our Go Direct™ Melt Station connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Caution: This product can expose you to chemicals, including nickel, which are known to the State of California to cause cancer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty