Would you like to log in again?
Go Direct™ Motion Detector
Supplier: VERNIER SOFTWARE & TECH SE
GDX-MD
470166-516EA 123.99 USD InStock
470166-516
Go Direct™ Motion Detector
Data Loggers
The Go Direct Motion Detector uses ultrasound to measure the position, velocity, and acceleration of moving objects.
  • The Go Direct Motion Detector can be used in a variety of experiments
  • Study position, velocity, and acceleration of carts when mounted on a track
  • Match graphs created in Graphical Analysis 4 software
  • Analyze the effects of air resistance on falling coffee filters
  • Investigate simple harmonic motion by monitoring a mass on a spring
The Go Direct Motion Detector accurately tracks objects as close as 15 cm and as far away as 2.5 m. Its built-in temperature compensation automatically adjusts for the difference in the speed of sound in cold and warm locations. Its unique design protects the sensor from falling objects during simple harmonic motion or free fall experiments.
Delivery information: Includes Vernier Go Direct motion detector, micro USB cable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty