Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Go Direct™ Electrode Amplifier

Supplier: VERNIER SOFTWARE & TECH SE
GDX-EA
470166-518EA 78.99 USD InStock
470166-518
Go Direct™ Electrode Amplifier
Data Loggers
The Go Direct Electrode Amplifier is a mV and pH amplifier that connects to Vernier electrodes and compatible third-party BNC electrodes.

  • Units: pH or mV
  • Input range (mV): ±1000 mV
  • Input range (pH): 0 to 14 pH

The Go Direct Electrode Amplifier offers an affordable way to expand your experiment options. Its BNC connector provides the flexibility to attach and remove a Vernier electrode, such as the pH Electrode, Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode. You can explore pH one day and potassium concentrations the next.

Because the Go Direct Electrode Amplifier is compatible with third-party electrodes, students learn consistent calibration and handling techniques on one amplifier using one software program. When electrodes expire or become damaged you need only replace the electrode—lowering your overall equipment costs.

The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.

Our Go Direct™ Electrode Amplifier connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.

Delivery information: Includes Go Direct EKG sensor, a package of 100 disposable electrodes, micro USB cable.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty