Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Go Direct Colorimeter
Supplier: VERNIER SOFTWARE & TECH SE
GDX-COL
470166-530EA 128.99 USD InStock
470166-530
Go Direct Colorimeter
Data Loggers
The four-wavelength Go Direct Colorimeter measures absorbance or transmittance of a liquid sample.
  • Conduct Beer’s law investigations
  • Determine the concentration of unknown solutions
  • Study changes in concentration vs. time
  • Monitor reaction rates
Use this sensor to explore absorbance and percent transmittance in a variety of experiments including Beer’s law (absorbance vs. concentration) and kinetic studies (concentration vs. time). Students select between four wavelengths (430 nm, 470 nm, 565 nm, 635 nm) to set up their experiment.
Go Direct Colorimeter features one-step calibration for all four wavelengths. It’s as simple as inserting a cuvette of distilled water and pressing the Calibrate button.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Colorimeter connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty