Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Go Direct™ Conductivity
Supplier: VERNIER SOFTWARE & TECH SE
GDX-CON
470166-536EA 128.99 USD InStock
470166-536
Go Direct™ Conductivity
Data Loggers
The Go Direct Conductivity Probe determines the ionic content of an aqueous solution by measuring its electrical conductivity.
  • The Go Direct Conductivity Probe can be used in a variety of experiments
  • Demonstrate diffusion of ions through membranes
  • Investigate the difference between ionic and molecular compounds, strong and weak acids, or ionic compounds that yield different ratios of ions
  • Measure Total Dissolved Solids (TDS)
Go Direct Conductivity Probe determines the ionic content of an aqueous solution by measuring its electrical conductivity. It features a built-in temperature sensor to simultaneously read conductivity and temperature. Automatic temperature compensation allows students to calibrate the probe in the lab and then make measurements outdoors without temperature changes affecting data. This temperature compensation can be turned off to perform conductivity studies as a function of temperature.
The Go Direct Conductivity Probe has a range of 0 to 20000 μS/cm to provide optimal precision in any given range. An alternating current at its electrodes prevents polarization and electrolysis, reducing contamination of solutions. Its ABS-body graphite electrode resists the corrosion typical in metal electrodes.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Conductivity Probe connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Caution: They are designed for educational use. They are not recommended for any industrial, medical, or commercial process such as life support, patient diagnosis, control of a manufacturing process, or industrial testing of any kind.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty