Would you like to log in again?
Go Direct™ 3-Axis Magnetic Field Sensor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-3MG
470166-540EA 88.99 USD InStock
470166-540
Go Direct™ 3-Axis Magnetic Field Sensor
Data Loggers
The Go Direct 3-Axis Magnetic Field Sensor allows you to determine the magnitude and direction of the magnetic field at any point in space.
  • The Go Direct 3-Axis Magnetic Field Sensor can be used in a variety of experiments
  • Quantify the magnetic field strength of induced magnets
  • Determine the declination and inclination of the Earth’s magnetic field at your location using vector components
  • Investigate the relationship between magnetic field strength, coils per unit length, and current in the center of a solenoid
The Go Direct 3-Axis Magnetic Field Sensor measures the components of the magnetic field along three orthogonal axes. If desired, measure the field along only two axes, or even one axis, choosing the direction that is best for the experiment.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ 3-Axis Magnetic Field Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty