Would you like to log in again?
Go Direct™ Drop Counter
Supplier: VERNIER SOFTWARE & TECH SE
GDX-DC
470166-542EA 108.99 USD InStock
470166-542
Go Direct™ Drop Counter
Data Loggers
Precisely records the number of drops of titrant added during a titration and then automatically converts it to volume.
  • Accurate drop counting at rates up to 6 drops/s
  • Directly connects wirelessly via Bluetooth® or wired via USB to your platform
Conducting a titration has never been easier. Use Go Direct Drop Counter in conjunction with our other Go Direct sensors, such as Go Direct pH, Go Direct Conductivity, or Go Direct ORP, to perform acid-base, conductometric, or potentiometric titrations.
Designed with a wide drop-detecting area, Go Direct Drop Counter removes the need for precise burette alignment. We’ve also included a large sensor slot for conventional stick-style sensors and a smaller, adjustable slot for temperature probes. The adjustable clamp easily fits our Stir Station and most laboratory ring stands and lattices.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Drop Counter connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct drop counter, plastic reagent reservoir, plastic valve with a double stopcock fitting, Vernier microstirrer (a magnetic stirrer that fits on the tip of your sensor), micro USB cable.
Caution: These products are designed for educational use. Our products are not designed nor are they recommended for any industrial, medical, or commercial process such as life support, patient diagnosis, control of a manufacturing process, or industrial testing of any kind.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty