Would you like to log in again?
Go Direct™ Temperature Probe
Supplier: VERNIER SOFTWARE & TECH SE
GDX-TMP GDX-TMP-TP
470166-546EA 77.99 USD InStock
470166-546 470166-526
Go Direct™ Temperature Probe
Data Loggers
A rugged, general-purpose temperature sensor that students can use to monitor temperature.
  • Conduct endothermic and exothermic reactions
  • Investigate the freezing and melting of water
  • Measure the energy content of foods
  • Examine the absorption of radiant energy
  • Monitor environmental conditions
The rugged, all-purpose Go Direct Temperature Probe gives students the advantage of go-anywhere, work-anywhere connectivity. Because it is also waterproof, Go Direct Temperature is more durable than sensors that are simply water resistant.
Unlike a traditional thermometer, Go Direct Temperature allows students to collect real-time temperature measurements of a single instance or over a period of time. Its temperature range and wireless capability make Go Direct Temperature the go-to sensor for real-world applications.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ Temperature Probe connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Go Direct temperature includes GO Direct temperature probe, micro USB cable, quick-start guide, Go Direct temperature teacher pack includes eight Go Direct temperature probes, charge station, eight micro USB cables, quick-start guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty