Would you like to log in again?
Go Direct™ pH Sensor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-PH GDX-PH-TP
470166-524PK 932.99 USD InStock
470166-524 470166-548
Go Direct™ pH Sensor
Data Loggers
A general-purpose pH sensor used to monitor pH of aqueous solutions.
  • Conduct acid-base titrations
  • Monitor pH change during chemical reactions
  • Test the pH and alkalinity of bodies of water
  • Investigate household acids and bases
The Go Direct pH Sensor is an important and versatile sensor for lab and field activities alike. It gives students the freedom to explore pH without the inconvenience of wires—no more spilled solutions or dangling wires. The Go Direct pH transmits live readings and captures data in real-time.
This sensor is comprised of a pH electrode connected to a Go Direct Electrode Amplifier via a BNC connector. This design enables you to use the Go Direct Electrode Amplifier with other Vernier electrodes, such as the Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ pH Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Go Direct pH sensor includes Go Direct Electrode amplifier,pH storage solution bottle with pH storage solution, micro USB cable, quick-start guide.
Go Direct pH Teacher Pack includes eight Go Direct pH Sensors, eight pH storage solution bottles with pH storage solution, charge station, eight micro USB cables, Quick-start guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty