Would you like to log in again?
Garage Physics Projectile Motion Sling Shot, Eisco Scientific
Supplier: Eisco Scientific
GP00014
470168-166EA 103 USD InStock
470168-166
Garage Physics Projectile Motion Sling Shot, Eisco Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Projectile motion is one of the most important topics in Newtonian mechanics, next to Hooke's Law for springs. Explore both in a very simple and robust setup.
  • This kit allows students to explore projectile motion writ-large with a rubber-band slingshot!
  • The material is high quality precision-cut Birch veneer pure bond plywood
  • All necessary parts are included and shown in picture
  • Made in the USA
By recording the time-of-flight of the ping pong ball, students can deduce the initial velocity for various rubber band settings. This can be used to experimentally determine the spring constant of the common rubber band in a dynamic setting.
By tipping the slingshot and breaking the initial velocity into its horizontal and vertical components, range can be predicted and measured. Range with 2 rubber bands is more than 30 feet. Additional rubber bands can launch the ping pong ball (or other projectiles) much farther.
Delivery information: Kit comes with a step-by-step educational guide and includes the slingshot, ping pong ball, and angled supports.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty