Would you like to log in again?
Garage Physics MURLIN Trebuchet, Eisco Scientific
Supplier: Eisco Scientific
GP00015
470168-168EA 159.6 USD InStock
470168-168
Garage Physics MURLIN Trebuchet, Eisco Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Who knew that the trebuchet, one of the most elegant of all medieval weapons ever invented, could be improved!
  • Explore how the use of a simple machine (pulley) efficiently redirects gravitational potential energy to throw objects farther than a traditional trebuchet
  • Manufactured with high quality, precision-cut Birch veneer pure bond plywood
  • Includes the trebuchet, ping pong ball, and 1 kg counterweight
By redirecting the pulling force of the counterweight with a pulley, the Multi-Radial Linear Node (MURLIN) Trebuchet is able to transfer much more energy to the projectile than the standard trebuchet. Assembled kit throws the included ping pong ball 30 feet or more.
In fact, this system, with only half the throwing arm of our standard trebuchet is able to throw just as far, if not farther. The included guide provides clear assembly instructions with pictures. It also contains pedagogy for exploring concepts of energy transfer using the scientific method.
Ordering information: A small Phillip's screwdriver is required for assembly.
Delivery information: Kit comes with a step-by-step educational guide.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty