Would you like to log in again?
Tri Pour Beakers
Supplier: DYN-A-MED PRODUCTS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
80096 80092 80094 80096 80091 80093 80098 80093
470174-586CS 70.7 USD InStock
470174-586 470174-585 470174-590 490012-442 470174-582 470174-581 470174-588 490012-439
Tri Pour Beakers
Beakers
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
  • Accurate Graduations to ±5
  • Tri-Cornered Lip for Dripless Pouring
  • Unbreakable
  • Resistant to Common Acids, Solvents, and Alkalis
  • Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty