Would you like to log in again?
Waterwise 3200 Countertop Distiller
Supplier: WATERWISE, INC. SE
Ward's®
8606 3212 1003 3250
470303-238EA 60.75 USD InStock
470303-238 470143-280 470303-164 470174-874
Waterwise 3200 Countertop Distiller
Distillation Systems Automatic Distillation Systems
Simple, one touch operation.
  • One-year limited warranty
  • No assembly, installation or hookups necessary
You'll love using the 3200. All you do is fill the stainless steel boiler with tap water, press the reset button, and soon you will have the purest water possible. After running a one-gallon cycle, keep a carafe ready for use later. As with all Waterwise distillers, the 3200 combines steam distillation with carbon post-filtration, resulting in optimum water purity.
Accessories information: Replacement filter, item 470174-874.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty