Would you like to log in again?
Plant Light and Water Tester
Supplier: LUSTER LEAF PRODUCTS INC SE
1830-A
470175-074CS 220.7 USD InStock
470175-074
Plant Light and Water Tester
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
A slender probe gauges the soil mixture at the roots of a plant, while a light sensor on the front of the unit measures the amount of ambient light.
  • Size: 4L×2½W×1½"H
  • Probe length: 6"
The light-weight unit needs no batteries, so there is nothing to replace or wear out. It includes a booklet with information on light and water requirements for over 150 plants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty