Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Coplin Staining Jars
Supplier: United Scientific Supplies
62101-PK/12 674
470174-652CS 125 USD InStock
470174-652 470175-194
Coplin Staining Jars
Jars Staining Jars
Coplin Jar with Screw-on Top Prevents Spills.
  • Quality slide making supplies
  • Slide staining tools
The screw-on cover prevents evaporation and contamination when staining. Each jar holds five single slides or ten back to back.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty