Would you like to log in again?
Artemia (Brine Shrimp) Hatchery
Supplier: Avantor
WARD470175-698
470175-698KT 133.99 USD InStock
470175-698
Artemia (Brine Shrimp) Hatchery
Educational Classroom Kits and Activities
Learn how to care for brine shrimp. Students build a brine shrimp hatchery of their own design and analyze their effectiveness through hatch rates and by using population sampling techniques. It is up to them to determine the optimal environmental conditions and the most efficient sampling procedure. Using the results of their first attempt, students can then develop a hypothesis that will improve their design.
  • Can also be used with triops or daphnia
  • Grades 6 - 12
  • Developed by Alan Nakagawa
Ordering information: Includes instruction manuals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty