Would you like to log in again?
Coffee Plant
Supplier: CHASE GREENHOUSES, INC.
867013
470176-168EA 12.3 USD InStock
470176-168
Coffee Plant
Specimens Live Specimens Plants - Living Material
Coffea arabica. Dicot. An attractive foliage plant as well as a productive one, this native of the tropics will grow easily in the classroom if placed in rich soil. The plant’s blossoms turn to bright red berries, each of which yield two coffee beans. It is supplied in a pot with care instructions. Size: 4–6".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty