Would you like to log in again?
Ward's® Freshwater Copepods
Supplier: VWR
WARD470176-228
470176-228EA 12.95 USD InStock
470176-228
Ward's® Freshwater Copepods
Specimens Live Specimens Aquaria - Living Material
The culture supplied includes copepods in various stages of development.
Cyclops or similar genus. Copepods molt numerous times in the nauplius and copepodid stages before reaching adulthood.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty