Would you like to log in again?
Banana Plug Patch Cords
Supplier: PRECISION MOULDED POLYMERS LIMITED
1350302190002 1350302150002 1350302150001 1350302190001
470176-240EA 8.05 USD InStock
470176-240 470176-242 470188-740 470188-742
Banana Plug Patch Cords
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Double ended banana plug jumper wires.
  • Available in red or black
  • 15 cm (6") or 61 cm (24") long
Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty