Would you like to log in again?
Algae Types Slides
Supplier: Avantor
WARD470176-244
470176-244KT 69.99 USD InStock
470176-244
Algae Types Slides
Slides Prepared Slides
Varying examples of algae classifications
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
This selection of slides is designed to supplement lessons found in most high school and introductory college textbooks. The representative selection allows students to study the classification of algae - a controversial topic due to the fact that algae may be grouped in as many as three separate kingdoms. All slides are whole mounts except for Fucus monoecious, which is a section.
Ordering information: Set includes Chlamydomonas(wm); Cladophora(wm); Desmids(wm); Oedogonium(wm); Pediastrum(wm); Spirogyra conjugation(wm); Blue-green algae(wm); Batrachospermum(wm); Volvox(wm); Euglena(wm); Diatoms(wm); Fucus monoecious (sec).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty