Would you like to log in again?
Mimosa
Supplier: CHASE GREENHOUSES, INC.
867100
470176-284PK 12.7 USD InStock
470176-284
Mimosa
Specimens Live Specimens Plants - Living Material
Mimosa pudica. Dicot. Also known as a sensitive plant, it is an ideal illustration of thigmotropism, or response to touch. The plant's leaves close when touched, and wilt or even drop off under more severe conditions but recovers quickly when the stimulus is taken away. It is native to tropical and subtropical regions of the Americas and prefers a moist environment. It is supplied in a pot with care instructions. Size: 4–8".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty