Would you like to log in again?
Ward's® Live Terrestrial Isopods (Pill Bugs and Sow Bugs)
Supplier: VWR
Avg. Ratings: 4.5 (6 Reviews)
REVIEWS (6)
Total Ratings: 6
Avg. Ratings: out of 5
Sort all by:
WARD470313-206 WACP470189-262 WARD470180-312 WARD470191-112 WARD470313-208 WARD470309-752 WARD470176-302 WARD470180-310 WARD470191-048
470309-752PK 79.95 USD InStock
470309-752 470180-310 470180-312 470313-208 470313-206 470191-112 470189-262 470191-048 470176-302
Ward's® Live Terrestrial Isopods (Pill Bugs and Sow Bugs)
Specimens Preserved Specimens Preserved Animal Materials
Often mistaken for beetles, these wingless crustaceans live their whole lives on land, feeding on decaying organic matter.
  • Easy to maintain in the laboratory or classroom
  • Do not bite or sting
  • Pill and Sow Bugs are crustaceans
  • Demonstrate differences in defensive behaviors between the two specimens
Pill Bugs (Armadillium vulgare) roll themselves into a ball if agitated.
Sow Bugs (Oniscus asellus) can be identified by their posterior "tail like" appendages, and will play dead when agitated. They are unable to roll into a ball.
Ordering information: These products contain living or perishable materials. Both Pill Bugs and Sow Bugs are easy to maintain in the laboratory or classroom. They prefer dark, damp locations such as under logs, leaves or rocks, and are most active at night. They are detritivores, and feed mostly on decaying organic matter.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (6)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty