Would you like to log in again?
Ward's® Live Green Hydra (Chlorohydra viridissima)
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470176-324 WARD470176-322
470176-322EA 15.25 USD InStock
470176-322 470176-324
Ward's® Live Green Hydra (Chlorohydra viridissima)
Specimens Live Specimens Animals - Living Material
Hydra are tiny animals closely related to jellyfish. They can be carnivorous as well as photosyntheic due to their symbiotic relationship with green algae Zoochlorellae.
  • Watch the feeding reflex in real-time
  • Observe the reproductive process of budding
  • Green hydra are smaller than the strictly carnivorous, Brown Hydra (also available)
Hydra have a sack-shaped body with a ring of tentacles around the mouth at the top. They use those tentacles, covered with paralyzing nematocysts, to trap small cruscaceans, stun them and bring them into the mouth where they are digested.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty