Would you like to log in again?
Ward's® Live Volvox aureus Culture
Supplier: VWR
WARD470176-362 WARD470176-360
470176-360EA 9.5 USD InStock
470176-360 470176-362
Ward's® Live Volvox aureus Culture
Specimens Live Specimens Protists - Living Material
Volvox aureus is a species of colonial green algae that forms spherical colonies of cells.
  • Extremely easy to visualize
  • Beautiful spherical, hollow colonies containing immature daughter cells
  • Volvox has green plastids and cell walls composed of cellulose
  • Cytoplasmic connections are the size of flagella- thinner than closely related V.globator (also available)
Volvox aureus is a species of Chlorophyte algae that can be found in freshwater lakes and ponds. When in optimal conditions, V. aureus will reproduce asexually; during times of drought or other adversity it will switch to sexual reproduction.
Genus: Volvox Species: aureus
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty