Would you like to log in again?
Ward's® Live Arcella Culture
Supplier: VWR
WARD470176-436
470176-436EA 9.5 USD InStock
470176-436
Ward's® Live Arcella Culture
Specimens Live Specimens Protists - Living Material
Usually found in freshwater and mosses, this organism has a hard umbrella shaped shell or "test", out of which finger-like pseudopodia project.
  • Contains a "Test"
  • Pseudopodia
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery information: Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty