Would you like to log in again?
Ward's® Philodina
Supplier: VWR
WARD470176-460
470176-460EA 9.5 USD InStock
470176-460
Ward's® Philodina
Specimens Live Specimens Aquaria - Living Material
Philodina sp. Easy to culture and easy to examine under a microscope because of the lack of a lorica (shell), these typical rotifers are perfect for general study. Culture instructions are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty