Would you like to log in again?
Mixed Flagellates
Supplier: Avantor
470176-466PK 13.95 USD InStock
470176-466 470176-468
Mixed Flagellates
Specimens Live Specimens Protists - Living Material
Specially Prepared “Real-Life” Combinations
The world is teeming with a wide variety of microlife, making studying samples of pond water complicated especially for the beginning student. To lessen the confusion, WARD’S has prepared different culture mixes that reflect protist combinations that occur in the real world, yet eliminates the hundreds of other life-forms found in a collected sample. Comes with a dichotomous key and either the Working with Algae or the Working with Protozoa manual. Supplied in a single jar.
Contents: Chlamydomonas, Eudorina, Euglena, Gonium, Pandorina, and Volvox.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty