Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ward's® Live Brown and Green Hydra Mixture
Supplier: VWR
WARD470176-480 WARD470176-482
470176-480EA 13.25 USD InStock
470176-480 470176-482
Ward's® Live Brown and Green Hydra Mixture
Specimens Live Specimens Animals - Living Material
You will receive a sample of both green and brown hydra mixed together, allowing you to observe the differences between the two organisms.
  • Watch the feeding reflex in real-time
  • Observe the reproductive process of budding
  • Look for similarities and differences between Brown hydra and green hydra
  • Discover which is more successful in a mixed culture
Hydra are tiny animals closely related to jellyfish. They can be found in freshwater river and streams where they attach themselves to plants and rocks.
Hydra have a sack-shaped body with a ring of tentacles around the mouth at the top. They use those tentacles, covered with paralyzing nematocysts, to trap small cruscaceans, stun them and bring them into the mouth where they are digested.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty