Would you like to log in again?
Ward's® Live Kocuria rhizophila (Micrococcus luteus) Culture
Supplier: VWR
WARD470176-534 WARD470176-536
470176-534EA 13.25 USD InStock
470176-534 470176-536
Ward's® Live Kocuria rhizophila (Micrococcus luteus) Culture
Specimens Live Specimens Bacteria - Living Material
Kocuria rhizophila is also commonly known by the name Micrococcus luteus. This organism is commonly isolated from soil, and is frequently used to test antimicrobial efficacy in the food industry.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Morphology: Cocci in tetrads
  • Motility: Negative
  • Respiration: Aerobic
  • Chromogenesis: Yes (yellow)
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ordering information: Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals. Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.
Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Caution: Though this organism is not considered to be pathogenic, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty