Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Live Serratia marcescens D1 Culture

Supplier: VWR
Avg. Ratings: 5.0 (1 Review)

REVIEWS (1)

Total Ratings: 1
Avg. Ratings: out of 5

Sort all by:

WARD470176-538 WARD470176-540
470176-538EA 13.25 USD InStock
470176-538 470176-540
Ward's® Live Serratia marcescens D1 Culture
Specimens Live Specimens Bacteria - Living Material
This organism is commonly isolated from soil, and depending upon the temperature of its environment, can produce a distinctive red pigment.

  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Negative
  • Morphology: Rods
  • Motility: Positive
  • Respiration: Aerobic
  • Chromogenesis: Temperature dependent (red at 25°C, colorless at 37°C )

All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.

Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ordering information: Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals. Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.

Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.

Caution: Though this organism is not considered to be pathogenic, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (1)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty